Slide wallets, FiloSlide™
Katalog # 720-2347
Supplier: CELLPATH
Specifications
- B×D:260×380 mm
- Färg:Genomskinlig
- Beskrivning:6 förpackningar (skåp+ 150 fack)
- Förp.:1
Specifications
About this item
These slide folders feature 21 compartments for 76×26, 50×76 and 76×76 mm slides.
- Hanging system
- Storage in conventional filing cabinets