Mobile concentrator system, CentriVap®
Leverantör: LABCONCO
Certifikat
Om denna produkt
This mobile system with or without heat boost combines the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12 to 13 mm rotor and solvent trap are also included. The interior can accommodate a vacuum pump — the only additional component necessary to begin processing.
- Microprocessor-controlled centrifugal motion up to 1725 rpm eliminates bumping and foaming
- Quick-Stop™ System:Once run time has expired or the RUN/STOP button is pressed, within seconds the vacuum pump stops, the valve opens to bleed air into the chamber, and the rotor stops
- Automatic vacuum release valve prevents loss of sample by releasing the vacuum before the rotor stops and during power failures
- Lid latch with safety sensor locks the lid in place when the rotor is turning and prevents start up when the lid is open
- Memory stores 9 user-set programs with parameters of temperature, heat time and run time; three Quick-Start™ buttons
- Collector drain speeds clean up and eliminate need for glass insert with non-corrosives
Optional: With microprocessor-controlled 300-watt heater that speeds evaporation by supplying a controlled amount of heat from OFF up to a maximum of 100° C in one degree increments.
Ordering information: Required accessories: Diaphragm vacuum pump or rotary vane direct drive vacuum pump, sample tubes.
Delivery: Supplied with rotor, built-in chemical trap canister and solvent trap insert.