Arsenazo III, Millipore®
Catalog # 1.10107.0005
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Molar Mass:776.36 g/mol
- Bulk Density:850 kg/m3
- pH:2.3 (10 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
About this item
:
:776.38 g/mol
ADR:6.1,
:M-Clarity™ quality level = MQ200
EINECS:216-788-6
UN:3465
:GHS06
:GHS09
Storage Temperature:Omgivande
CAS Number:1668-00-4
Formula:C₂₂H₁₈As₂N₄O₁₄S₂
MDL Number:MFCD00036695