Tributyl acetylcitrate
Catalog # PROOMM0499.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:500 mg
- Seq:2725
Specifications
About this item
:
:402.49 g/mol
EINECS:201-067-0
CAS Number:77-90-7
Formula:C₂₀H₃₄O₈
MDL Number:MFCD00043554