Microscope slide for LCP
LCP-sandwichset, 128×85 (slide) mm
112×77 (cover slip) mm
112×77 (cover slip) mm
Katalog # MARI0890003
Leverantör: MARIENFELD
Gå till:
Product Details & Documents
The lipidic cubic phase (LCP) sandwich set consists of a base slide and a cover slip and is optimized for the crystallographic study of membrane proteins.
- Surface with hydrophobic glass
- Optimised for brightfield, UV and fluorescent microscopy
- Can be used in G protein–coupled receptors (GPCR) structural biology
Specifikationer
- Beskrivning:LCP-sandwichset
- L×B:128×85 (slide) mm
112×77 (cover slip) mm