Banded earplugs, QB2HYG®
Catalog # 112-2916
Leverantör: HONEYWELL SAFETY
Specifikationer
- Färg:Orange/röd
- Beskrivning:Supra-aural banded earplugs, QB2HYG®
- Förp.:100 PAIR
Specifikationer
Om denna produkt
These foam earplugs offer solid protection with supra-aural fit for people who are exposed to intermittent noise.
- Smooth, ergonomic pods fit inside the opening of the ear canal for maximum protection
- Bright colours for improved visibility
- Lightweight and portable
SNR: 25 dB
Certifications: EN 352-2
Packaging: In a resealable bag.