Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Flaskor
Sprutflaskor
Wash bottles, narrow neck, with screw cap, BOLA
Wash bottles, narrow neck, with screw cap, BOLA
Wash bottles, narrow neck, with screw cap, BOLA
Leverantör:
Bohlender
Certifikat
Expandera bilden
Wash bottles, narrow neck, with screw cap, BOLA
Leverantör:
Bohlender
Om denna produkt
PFA, translucent.
Temperature resistance: from −200 to +250 °C
Universal chemical resistance
Non porous
Graduated