Teststrips, Test av total hårdhet, Reflectoquant®, Supelco®
Catalog # 1.16997.0001
Supplier: Merck
CAS Number:
Specifications
- Kemisk metod:Phthalein complexone
- Förp.:50 Tests
- Beskrivning:Test av total hårdhet
- M-Clarity quality level:MQ100
- Mätområde:0,1 - 30,0 °d (1 - 300 mg/l CaO)
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly