Teststrips, Askorbinsyratest, Reflectoquant®, Supelco®
Catalog # 1.16981.0001
Supplier: Merck
CAS Number:
Specifications
- Kemisk metod:Phosphormolybdenum blue
- Förp.:50 Tests
- Beskrivning:Askorbinsyratest
- M-Clarity quality level:MQ100
- Mätområde:25 - 450 mg/l C₆H₈O₆
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly