Sporicide, Minncare® Cold Sterilant
Katalog # 148-0062
Leverantör: MARCOR PURIFICATION
Specifikationer
- Beskrivning:Minncare® Cold Sterilant
- Förp.:5 l
Specifikationer
Om denna produkt
Minncare® Cold Sterilant concentrate is an effective viricide, bactericide, fungicide, and sporicide for surface disinfection in pharmaceutical cleanroom environments. It is completely and contains only pharmaceutical grade raw materials.
- Leaves no measurable air residue following ventilation of room
- Faster, safer, and less corrosive than aldehyde-based liquids
- EPA registered for fogging as part of normal cleaning and disinfecting procedures