Kiselgel, Pore size: 100 Å 70-230 mesh, high purity for column chromatography, Supelco®
Catalog # 60745-1KG
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:1 kg
- Seq:4024
Specifications
About this item
:
:60.08 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Omgivande
CAS Number:112926-00-8
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232