1-Butoxi-2,3-epoxipropan
Catalog # 8.01553.0250
Supplier: Merck
CAS Number:
Specifications
- Förp.:250 mL
- Seq:2590
Specifications
Specification Test Results
- Solubility:20 g/l (20 °C)
- Molar Mass:130.19 g/mol
- Boiling Point:163 - 165 °C (1013 mbar)
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:43 °C
- Density:0.92 g/cm3 (20 °C)
- Ignition Point:215 °C
Specification Test Results
About this item
:
:130.19 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:219-376-4
UN:3271
:GHS02
:GHS08
:GHS07
Storage Temperature:Omgivande
CAS Number:2426-08-6
Formula:C₇H₁₄O₂
MDL Number:MFCD00005145