ContactUsLinkComponent
Diamond pen with wooden handle
Diamond pen with wooden handle
Katalog # MARI6120300
Leverantör:  MARIENFELD
undefined
Diamond pen with wooden handle
Katalog # MARI6120300
Leverantör:  MARIENFELD
Specifikationer
  • Beskrivning:
    Diamantpenna, trähandtag
  • Förp.:
    1
Specifikationer
Om denna produkt
For permanent labelling of microscope slides and other glass products. Indelibly engraved markings comply particularly with the guidelines of GLP.
  • Resistant to humidity, heat, cold and solvents
  • With wooden handle