Natriumklorid
Catalog # USBIS5000-1-1KG
Supplier: US Biological
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle for solids
- Förp.:1 kg
- Seq:4024
Specifications
About this item
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2,16 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477