Kaliumklorid
Catalog # PROONCSGC76208
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:50 g
- Seq:3368
Specifications
About this item
:
:74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1,98 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360