Litiummetaborat, Spectromelt® A20 for XRF (X-ray fluorescence), fused, Supelco®
Katalog # 1.12996.1000
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 kg
- Seq:4024
Specifications
About this item
:
:49.75 g/mol
Merck Index:13,05546
Boiling Pt:1719 °C (1013 hPa)
Melting Pt:845 °C
Density:1,4 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Omgivande
EINECS:236-631-5
CAS Number:13453-69-5
Formula:BLiO₂
MDL Number:MFCD00011089