Order Entry
Sweden
ContactUsLinkComponent
Saltsyra 0.01 mol/l (0.01 N)
Saltsyra 0.01 mol/l (0.01 N)
Catalog # FFCHFF9101000
Supplier:  FF CHEMICALS
CAS Number:  
Saltsyra 0.01 mol/l (0.01 N)
Catalog # FFCHFF9101000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Förpackningstyp:
    Glass bottle
  • Förp.:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324