Nonylbenzene
Catalog # EHERC15628000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:0,1 g
- Seq:3020
Specifications
About this item
:
:204.36 g/mol
Boiling Pt:282 °C (1013 hPa)
Melting Pt:–24 °C
Density:0,8584 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:214-103-5
CAS Number:1081-77-2
Formula:C₁₅H₂₄
MDL Number:MFCD00009575