Containers, multi-purpose, with screw caps
120 ml, Sterila, Röd, individuellt förpackade, kartong à 250
Leverantör: RATIOLAB
Gå till:
Product Details & Documents
Made of PP with red screw-cap.
- Multi-purpose
- Ideal for the clinical routine
- Good chemical and temperature resistance
Suitable for sampling, transport and storage of solid, pasty and liquid samples.
Specifikationer
- Förp.:individuellt förpackade, kartong à 250
- Typ:Sterila
- Kapacitet:120 ml
- Seq:02
- Lock, färg:Röd