Transport trolleys
Katalog # BOCH17330
Leverantör: BOCHEM
Specifikationer
- Beskrivning:Transport cart, ROLLO
- B×D×H:603×403×950 mm
- Förp.:1
- Antal hyllor:for 2 baskets
Specifikationer
Om denna produkt
Transport trolley made of electrolytically polished stainless steel.
- With handle and four guide rolls (two lockable).
Ordering information: Please order baskets separately.