
4-Tolualdehyde för syntes, Sigma-Aldrich®
Catalog # 8.06179.0250
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:0.25 g/l (25 °C)
- Melting Point:-6 °C
- Molar Mass:120.15 g/mol
- Boiling Point:204 °C (1013 hPa)
- Vapor Pressure:0.33 hPa (25 °C)
- Flash Point:71.5 °C DIN 51758
- Refractive Index:1.5447 (20 °C, 589 nm)
- Explosion Limit:0.9 - 5.6 %(V)
- Density:1.02 g/cm3 (20 °C)
- Ignition Point:395 °C DIN 51794
- Synonyms:p-Tolualdehyde, p-Toluylaldehyde
- Hill Formula:C8H8O
- Chemical Formula:4-(CH3)C6H4CHO
Specification Test Results
About this item
:
:120.15 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–6 °C
Density:1,02 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:72 °C
Storage Temperature:Omgivande
EINECS:203-246-9
:GHS07
CAS Number:104-87-0
Formula:C₈H₈O
MDL Number:MFCD00006954