Pipette filler
Katalog # KART198
Leverantör: KARTELL
Specifikationer
- Kapacitet:10 ml
- Beskrivning:Pipettfyllare med kolv, ABS/HDPE, med graderingar
- Förp.:1
Specifikationer
Om denna produkt
Pipette filler with plunger, will handle pipettes up to 10 ml.
- Comfortable grip
- Simply press plunger and release to fill pipette
- Knurled cap adjusts the plunger stroke for repeating pre-set volume
- Rubber chuck will hold pipette securely and safely
- Graduations on the body
- Moulded in yellow ABS