Skopa
med stav, 1000 ml, 115 cm
331-0056
: BURKLE
Stainless steel V2A (1.4301).
- Complete with rod
- No openings, undercuts or crevices in which residues could collect
- Pore-free finish produced by high quality laser welding seams
- Sterilisable by flame or steam sterilisation
- Typ:med stav
- Kapacitet:1000 ml
- Längd:115 cm