Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Elektrokemimätare
pH-mätare
Accessories for Quadroline
Accessories for Quadroline
Accessories for Quadroline
Catalog # 662-0281
Supplier:
WTW ONLINE
Expand image
Accessories for Quadroline
Catalog # 662-0281
Supplier:
WTW ONLINE
Specifications
Beskrivning:
pH/ORP sensor SensoLyt® 700-7, with integrated preamplifier and NTC; cable length 7 m
Förp.:
1
För:
EcoLine and QuadroLine® monitors
Specifications
About this item
Product Details
Product Details