Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Brickor/Skålar/Burkar
Avdunstningsskålar
Evaporating dishes
Evaporating dishes
Evaporating dishes
Leverantör:
RSG
Certifikat
Expandera bilden
Evaporating dishes
Leverantör:
RSG
Om denna produkt
Made of stainless steel 1,4301.
Very good temperature resistance
Very good chemical resistance
Easy to clean