Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Brickor/Skålar/Burkar
Laboratoriebrickor/skålar
Bowls
Bowls
Bowls
Leverantör:
BURKLE
Certifikat
Expandera bilden
Bowls
Leverantör:
BURKLE
Om denna produkt
Made of PP, white.
With widce holding edge
Can be stacked
Sterilisable up to +121 °C
Particularly stable bowl.