Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern, Class B
Automatic burettes, Schilling pattern, Class B
Supplier:   KARTELL
Automatic burettes, Schilling pattern, Class B
Supplier:  KARTELL
Certificates
Automatic burettes, Schilling pattern, Class B
Automatic burettes, Schilling pattern, Class B
2 Options Available Below
Product Details & Documents
PMP/PE, class B.
  • Push button for drop-by- drop delivery
  • PE reservoir with stable LDPE base
According to DIN 12.700 standards, class B
Certifications: According to DIN 12.700 standards, class B
Product Family Options
Product Information
  • Kapacitet
    Indelning
    Availability
    Price
  • 25 ml
    0,1 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3 425,00 kr
    3 425,00 kr
    1 * 1 Items
       3 425,00 kr
  • 50 ml
    0,1 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3 600,00 kr
    3 600,00 kr
    1 * 1 Items
       3 600,00 kr