Cleanroom floor wipes, sterile, AlphaLite® II
Catalog # 115-1893
Leverantör: Texwipe
Specifikationer
- Förp.:10 torkdukar/påse, 1 innerpåse med 10 torkdukar, 34 påsar/låda
- Beskrivning:TX3424
- L×B:600×250 mm
Specifikationer
Om denna produkt
Cut-edge polyester single ply wipe that is ideal for floor cleaning and for applying cleaning solutions and disinfectants on floors.
- Large size for improved application on floor surfaces
- Size is compatible with most mopping systems
- Improved contamination control
Suitable for use in Class 100, ISO 5 and Class 1000, ISO 6 environments.
Packaging: Packaging is adapted for sterile and highly critical environments.