Jars for organ storage, DURAN®
Katalog # SCOT242042409
Leverantör: DWK Life Sciences
Specifikationer
- Ø utv.:54 mm
- Kapacitet:75 ml
- Höjd:75 mm
- Förp.:10
Specifikationer
Om denna produkt
DURAN®, borosilicate glass 3.3, clear. Without stopper.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Organ storage jars, cylindrical vessels with a wide neck, are used to store materials.