ContactUsLinkComponent
Bägare, låg modell, PFA
Bägare, låg modell, PFA
Catalog # KART1538
Supplier:  KARTELL
Supplier #: 1538
undefined
Bägare, låg modell, PFA
Catalog # KART1538
Supplier:  KARTELL
Supplier #: 1538
Log in for availability
Select an Option
88 mm
Select an Option
100 ml
Select an Option
500 ml
Select an Option
122 mm
Select an Option
05
Select an Option
1
About this item
PFA, translucent, with spout.
  • Space-saving stackable design
  • High thermal resistance from −250 to +270 °C
  • Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.