Cleanroom floor wipes, AlphaWipe®
: Texwipe
These mop covers are constructed from 100% continuous-filament polyester in a double-knit, no-run interlock pattern.
- Ultra low levels of particles and extractables
- Continuous-filament fiber prevents shedding
- Prevents cleanroom contamination
- Resists abrasion with rigorous use or rough surfaces
- Excellent chemical resistance
Cleanroom laundered and packaged, AlphaWipe® is an ultraclean and highly sorbent wipe ideal for critical environments. They are designed for use with Doodleduster™ mops by 3M™ but can also be used as a work surface cover. Ideal for mopping in sensitive areas, such as integrated circuit fabrication facilities.
Suitable for use in ISO class 3 - 4 cleanrooms.
Colour: White
: Cleanroom laundered and packaged.