Bägare, Berzelius-typ, DURAN®
Catalog # SCOT211260106
Supplier: DWK Life Sciences
Specifications
- Ø utv.:50 mm
- Kapacitet:100 ml
- Höjd:78 mm
- Förp.:1
Specifications
About this item
DURAN® borosilicate glass 3.3, clear. Without spout and graduation.
- Very good chemical resistance
- High temperature resistance
- Autoclavable