Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lock
Dropplock
Closure for dropping bottles, 302 series
Closure for dropping bottles, 302 series
Closure for dropping bottles, 302 series
Catalog # 215-2471
Supplier:
Kautex
Expand image
Closure for dropping bottles, 302 series
Catalog # 215-2471
Supplier:
Kautex
Specifications
Gänga:
18 mm
Beskrivning:
Combination closure with screw-on dropping tip, HDPE
Förp.:
1
För flaskor:
10/25/50 ml
Specifications
About this item
Product Details
Documentation
Product Details
Documentation