Order Entry
Sweden
ContactUsLinkComponent
3,5-Dibromosalicylic acid
3,5-Dibromosalicylic acid
Catalog # MOLEM20609972
Supplier:  Molekula
CAS Number:  
3,5-Dibromosalicylic acid
Catalog # MOLEM20609972
Supplier:  Molekula
CAS Number:  

Specifications

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :295.91 g/mol
Merck Index:13,03053
Boiling Pt:365 °C (1013 hPa)
Melting Pt:228…229 °C
Storage Temperature:Omgivande
EINECS:221-570-9
 :GHS07
CAS Number:3147-55-5
Formula:C₇H₄Br₂O₃
MDL Number:MFCD00002441