ContactUsLinkComponent
Stopcocks for desiccators
Stopcocks for desiccators
Catalog # BRND82415
Supplier:  Brand
undefined
Stopcocks for desiccators
Catalog # BRND82415
Supplier:  Brand
Specifications
  • För:
    NOVUS
  • Beskrivning:
    Kran med nålventil, PTFE, kärnstorlek 24/29, öppning 0–4 mm
  • Förp.:
    1
Specifications
About this item
Stopcocks made of borosilicate glass 3.3 are suitable for Novus desiccators with lid socket.
  • Lateral venting helps to reduce turbulence
  • Nozzle with outer Ø 10 mm