Orders LinkContactUsLinkComponent
Adapters, bent, glass
Adapters, bent, glass
29/32 NS, 1×14/23+1×29/32 RN NS
Katalog # LENZ05313044
Leverantör:   Lenz Laborglas
Adapters, bent, glass
Katalog # LENZ05313044
Leverantör:  Lenz Laborglas
Adapters, bent, glass
Adapters, bent, glass
29/32 NS, 1×14/23+1×29/32 RN NS
Price
1 425,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
DURAN® Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Connector, bent, with cone and lateral socket NS 29/32, with socket NS 14/23 (for thermometer or air leak tube).
Dokument
Specifikationer
  • Kärna:
    29/32 NS
  • NS-hylsa:
    1×14/23+1×29/32 RN NS