Varningstejp, 766
Tejp, varningstext, svart/gul, vinylbaksida med gummi, 50,8 mm
Katalog # 819-0123
Leverantör: 3M
Gå till:
Produktdetaljer och dokument
These 766 barrier tapes are made of vinyl with a rubber adhesive and are ideal for hazard warning.
- Conformable backing ideal for use on curved and irregular surfaces
- Maintains vivid colour over time for long-lasting quality
- Rubber adhesive provides good instant adhesion on many surfaces
Used for marking of equipment pinch points or gears for warning purposes, temporary floor marking around equipment or spills.
Length: 33,0 m
Specifikationer
- Bredd:50,8 mm
- Beskrivning:Tejp, varningstext, svart/gul, vinylbaksida med gummi