Destillationsrör för ångdestillation
Katalog # LENZ05033044
Leverantör: Lenz Laborglas GmbH & CO.KG
Specifikationer
- Kärna:29/32 NS
- NS-hylsa:29/32 NS
- Förp.:1
Specifikationer
Om denna produkt
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With two cones NS 29/32 and one DIN thread GL 14.