4-Metyl-2-pentanon, EMPLURA®, Supelco®
Catalog # 8.20820.9190
Supplier: Merck
CAS Number:
Specifications
- Förp.:190 L
- Seq:5514
Specifications
About this item
:
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117,4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0,801 g/cm³ (20 °C)
Flash Pt:16 °C
EINECS:203-550-1
UN:1245
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Storage Temperature:Omgivande
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938