Dietyleter, EMPARTA® ACS for analysis, Supelco®
Catalog # 1.07026.2500
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:2,5 L
- Seq:5096
Specifications
About this item
:
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0,7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Omgivande
EINECS:200-467-2
UN:1155
ADR:3,I
:M-Clarity™ quality level = MQ200
REACH:01-2119535785-29
:GHS02
:GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646