Etylacetat 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77,1 °C (1013 hPa)
Density:0,902 g/cm³
Flash Pt:–4 °C
Storage Temperature:Omgivande
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Miljöcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Residual solvents:≤10 % MTBE