Pipette tray for drawer
BRND28305
: Brand
- För:cirka 30 graderade pipetter (1–20 ml)
- Beskrivning:Pipettbricka
- B×D×H:420×300×30 mm
- Förp.:1
Pipette tray made of PVC.
- Four compartments
- For approx. 30 graduated pipettes (1 to 20 ml)
Pipette tray made of PVC.