Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96-brunnars mikroplatta med U-botten, 300 μl
Katalog # 391-5708
Leverantör:   Brand
Microplates, 96-well
Katalog # 391-5708
Leverantör:  Brand
Microplates, 96-well
Microplates, 96-well
96-brunnars mikroplatta med U-botten, 300 μl
Price
1 825,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents
PP, round bottom wells, non sterile.
  • Compatible with virtually all leading microplate centrifuges
  • Raised well rims minimise cross-contamination
  • Plates can be sealed using self-adhesive films, such as DMSO-resistant sealing film (cross-cut) with alphanumeric coding
For volumes up to 300 µl.
Documents
Specifikationer
  • Beskrivning:
    96-brunnars mikroplatta med U-botten, 300 μl