Luteolin, Millipore®
Catalog # 440025-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:5 mg
- Seq:2210
Specifications
About this item
:
:286.24 g/mol
EINECS:207-741-0
:GHS07
CAS Number:491-70-3
Formula:C₁₅H₁₀O₆
MDL Number:MFCD00017309