Kalciumklorid dihydrat ≥99%, white solid, Molecular Biology Grade, Millipore®
Catalog # 208291-250
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle for solids
- Förp.:250 g
- Seq:3545
Specifications
About this item
:
:147.01 g/mol
Merck Index:13,01660
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1,85 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:233-140-8
:GHS07
CAS Number:10035-04-8
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613