Citronsyra monohydrat
Katalog # HACH2254232
Supplier: Hach
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:100 mL
- Seq:2440
Specifications
About this item
:
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1,552 g/cm³ (20 °C)
EINECS:201-069-1
:GHS07
Storage Temperature:Omgivande
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972