Räknekammare, BLAUBRAND® Neubauer Förbättrad
Katalog # 612-5713
Leverantör: Brand
Specifikationer
- Typ:Neubauer Improved
- Beskrivning:Counting chamber, without clips, bright-line
- Förp.:1
Specifikationer
Om denna produkt
This counting chamber feature the same ruling as improved Neubauer but with a rhodium coated chamber bottom.
- Rulings engraved into the rhodium layer appear bright under normal microscope settings
- By altering the contrast, the microscope image can be reversed
- Rulings can appear brighter or darker, as required