Natriumhydroxid 0.1 mol/l (0.1 N) i vattenlösning, AVS TITRINORM Reag. Ph. Eur. volumetrisk lösning
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1,003 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0998 to 0.1002 mol/l
- Conforms to BDH 19145:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Cubitainer (ZPACK8000467) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC31770.294 spec_VWRC31770.294 spec_VWRC31770.363 spec_VWRC31770.363 spec_VWRC31770.374
Product specification
More