Lock för PCR-platta med 96 brunnar, Corning® Thermowell®
Catalog # 734-1754
Supplier: Corning
Specifications
- Sterilitet:Icke sterilt
- Färg:Klar
- Beskrivning:Lock för PCR-platta med 96 brunnar, PC
- Förp.:25
Specifications
About this item
Polycarbonate lids are thermally stable up to 135 °C and can be used on Corning® Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays.
Lids are intended to be covers only and do not prevent evaporation.