Nalgene® Trattar med kort skaft, räfflade
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent.
- Shatter-proof
- Good chemical resistance
- Good temperature resistance
Reliable long-term tool for the lab.
Specifications
- Catalog No:
- NALG4262-0080
- NALG4262-0110
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)